Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanecarboxamide 97.0+%, TCI America™
SDP

Catalog No. A20781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20781G 1 g
A20785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20781G Supplier TCI America Supplier No. A20781G

Chemical Identifiers

CAS 5511-18-2
Molecular Formula C11H17NO
Molecular Weight (g/mol) 179.263
MDL Number MFCD00077200
InChI Key CKBZJTAMRPPVSR-UHFFFAOYSA-N
Synonym 1-adamantanecarboxamide, adamantanecarboxamide, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxamide, tricyclo 3.3.1.13,7 decane-1-carboxamide, 3r,5s,7s-adamantane-1-carboxamide, 2-carbamoyladamantane, pubchem8792, admantane-1-carboxamide, acmc-209ll3
PubChem CID 21686
IUPAC Name adamantane-1-carboxamide
SMILES C1C2CC3CC1CC(C2)(C3)C(=O)N

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 1 g
Formula Weight 179.26
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantanecarboxamide
RTECSNumber AU4452000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.