Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanemethanol, 98%

Catalog No. p-7027236
Click to view available options
Quantity:
5 g
25 g

CAS: 770-71-8 | C11H18O | 166.264 g/mol

1-Adamantanemethanol was used in the synthesis of axially disubstituted silicon(IV) phthalocyanines. It is used as a reagent in the synthesis of adamantane and noradamantane based histone deacetylase (HDAC) inhibitors for the treatment of cancer. It is also used as a reagent in the synthesis of (1-adamantyl)methyl glycidyl ether, a versatile building block for living polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanemethanol was used in the synthesis of axially disubstituted silicon(IV) phthalocyanines. It is used as a reagent in the synthesis of adamantane and noradamantane based histone deacetylase (HDAC) inhibitors for the treatment of cancer. It is also used as a reagent in the synthesis of (1-adamantyl)methyl glycidyl ether, a versatile building block for living polymerization.

Solubility
Insoluble in water. Solubility in methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 770-71-8
Molecular Formula C11H18O
Molecular Weight (g/mol) 166.264
MDL Number MFCD00074751
InChI Key MDVGOOIANLZFCP-UHFFFAOYSA-N
Synonym 1-adamantanemethanol, adamantan-1-ylmethanol, 1-adamantane methanol, 1-hydroxymethyl adamantane, adamantane-1-methanol, adamantan-1-yl-methanol, 1-hydroxymethyladamantane, tricyclo 3.3.1.1'3,7 dec-1-ylmethanol, adamantan-1-yl methanol, tricyclo 3.3.1.13,7 decanemethanol
PubChem CID 64556
IUPAC Name 1-adamantylmethanol
SMILES C1C2CC3CC1CC(C2)(C3)CO

Specifications

Melting Point 115°C to 118°C
Quantity 5 g
Beilstein 1853339
Solubility Information Insoluble in water. Solubility in methanol (almost transparency).
Formula Weight 166.26
Percent Purity 98%
Chemical Name or Material 1-Adamantanemethanol
EINECSNumber 212-225-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.