Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanemethanol, 99%

Catalog No. AC180050100
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle

CAS: 770-71-8 | C11H18O | 166.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 770-71-8
Molecular Formula C11H18O
Molecular Weight (g/mol) 166.26
MDL Number MFCD00074751
InChI Key MDVGOOIANLZFCP-UHFFFAOYSA-N
Synonym 1-adamantanemethanol, adamantan-1-ylmethanol, 1-adamantane methanol, 1-hydroxymethyl adamantane, adamantane-1-methanol, adamantan-1-yl-methanol, 1-hydroxymethyladamantane, tricyclo 3.3.1.1'3,7 dec-1-ylmethanol, adamantan-1-yl methanol, tricyclo 3.3.1.13,7 decanemethanol
PubChem CID 64556
IUPAC Name 1-adamantylmethanol
SMILES C1C2CC3CC1CC(C2)(C3)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 115.0°C to 118.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: .. Other solubilities: soluble in hexane and ethyl alcohol
Formula Weight 166.26
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantanemethanol, 99%
Show More Show Less
EINECSNumber 212-225-3
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.