Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanol 98.0+%, TCI America™
SDP

Catalog No. A0939250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0939250G 250 g
A093925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0939250G Supplier TCI America Supplier No. A0939250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 768-95-6
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00074729
InChI Key VLLNJDMHDJRNFK-UHFFFAOYSA-N
Synonym 1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol
PubChem CID 64152
IUPAC Name adamantan-1-ol
SMILES C1C2CC3CC1CC(C2)(C3)O

Specifications

Melting Point 280°C
Color White
Quantity 250 g
Formula Weight 152.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantanol
EINECSNumber (4)-1945
RTECSNumber AU4980000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.