Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantyl bromomethyl ketone, Thermo Scientific™

Catalog No. AC165150050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5122-82-7
Molecular Formula C12H17BrO
Molecular Weight (g/mol) 257.17
MDL Number MFCD00074740
InChI Key KWCDIRFSULAMOC-UHFFFAOYSA-N
Synonym 1-adamantyl bromomethyl ketone, 1-adamantan-1-yl-2-bromoethanone, 1-bromoacetyl adamantane, 1-1-adamantyl-2-bromoethanone, bromomethyl 1-adamantyl ketone, bromomethyl adamantyl ketone, ethanone, 2-bromo-1-tricyclo 3.3.1.1 sup 3,7 dec-1-yl, adamant-1-yl bromomethyl ketone, 1-1-adamantyl-2-bromoethan-1-one, 2-bromo-1-tricyclo 3.3.1.1 sup 3,7 dec-1-ylethanone
PubChem CID 197820
IUPAC Name 1-(adamantan-1-yl)-2-bromoethan-1-one
SMILES BrCC(=O)C12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 75°C to 79°C
Packaging Glass bottle
Quantity 5 g
Formula Weight 257.16
Chemical Name or Material 1-Adamantyl bromomethyl ketone
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 225-860-6
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.