Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Allyl-3-(2-hydroxyethyl)-2-thiourea 98.0+%, TCI America™
SDP

Catalog No. A0230500G
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 105-81-7
Molecular Formula C6H12N2OS
Molecular Weight (g/mol) 160.235
MDL Number MFCD00002838
InChI Key VUVPNTYTOUGMDG-UHFFFAOYSA-N
Synonym 1-allyl-3-2-hydroxyethyl-2-thiourea, 1-allyl-3-2-hydroxyethyl thiourea, thiourea, n-2-hydroxyethyl-n'-2-propenyl, n-allyl-n'-2-hydroxyethyl thiourea, n-2-hydroxyethyl-n'-allylthiourea, n-2-hydroxyethyl-n'-2-propenylthiourea, n-allyl-n'-2-hydroxyethylthiourea, urea, 1-allyl-3-2-hydroxyethyl-2-thio, n-allyl-n'-beta-hydroxyethyl thiourea
PubChem CID 2735273
IUPAC Name 1-(2-hydroxyethyl)-3-prop-2-enylthiourea
SMILES C=CCNC(=S)NCCO

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 500 g
Formula Weight 160.24
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Allyl-3-(2-hydroxyethyl)-2-thiourea
EINECSNumber (2)-1764
RTECSNumber YR7913000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.