Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich 1-Allyl-3-Methylimidazolium Chloride
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 g
50 g
Description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.AMIM-Cl may be used: As a solvent to dissolve cellulose during the preparation of regenerated cellulose materials.As a solvent for the acetylation of cornhusk cellulose to form cornhusk cellulose acetates (CCAs).As a plasticizer for cornstarch to form AMIM-Cl-plasticized starch film.
Specifications
Specifications
| CAS | 65039-10-3 |
| Molecular Formula | C7H11ClN2 |
| Linear Formula | C7H11ClN2 |
| MDL Number | MFCD07784450 |
| Quantity | 5 g |
| Synonym | AMIM-Cl |
| Molecular Weight (g/mol) | 158.63 g/mol |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction