Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-4-bromonaphthalene 98.0+%, TCI America™
SDP

Catalog No. A188625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A188625G 25 g
A18865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A188625G Supplier TCI America Supplier No. A188625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2298-07-9
Molecular Formula C10H8BrN
Molecular Weight (g/mol) 222.09
InChI Key LIUKLAQDPKYBCP-UHFFFAOYSA-N
Synonym 4-Bromo-1-naphthylamine
PubChem CID 75303
IUPAC Name 4-bromonaphthalen-1-amine
SMILES C1=CC=C2C(=C1)C(=CC=C2Br)N

Specifications

Melting Point 102°C
Color Purple-White
Quantity 25 g
Formula Weight 222.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-4-bromonaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.