Learn More
1-Amino-4-hydroxyanthraquinone, 96%
Description
The use of the inclusion complex of 1-amino-4-hydroxyanthraquinone (AHA) in the internal cavity of β-cyclodextrin confers increased sensitivity, selectivity and detection limit, and allows the determination of 10-70 ng ml-1 beryllium, compared to 60-500 ng ml-1 in the absence of the cyclodextrin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
The use of the inclusion complex of 1-amino-4-hydroxyanthraquinone (AHA) in the internal cavity of β-cyclodextrin confers increased sensitivity, selectivity and detection limit, and allows the determination of 10-70 ng ml-1 beryllium, compared to 60-500 ng ml-1 in the absence of the cyclodextrin.
Solubility
Soluble in water, ethanol, acetone, benzene.
Notes
Store at room temperature. Incompatible with oxidizing agents.
 
            Specifications
Specifications
| Melting Point | 207°C to 209°C | 
| Quantity | 25 g | 
| Solubility Information | Soluble in water,ethanol,acetone,benzene. | 
| Formula Weight | 239.23 | 
| Percent Purity | 96% | 
| Chemical Name or Material | 1-Amino-4-hydroxyanthraquinone | 
Safety and Handling
| Hazard Category | H304-H341-H351 | 
| Hazard Statement | GHS H Statement H341 Suspected of causing genetic defects. | 
| Precautionary Statement | P201-P202-P281-P301+P310-P308+P313-P331-P501c | 
| EINECSNumber | 204-163-0 | 
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
 
                                