Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-4-hydroxyanthraquinone 96%, Thermo Scientific™

Catalog No. p-56594
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 116-85-8
Molecular Formula C14H9NO3
Molecular Weight (g/mol) 239.23
MDL Number MFCD00001223
InChI Key AQXYVFBSOOBBQV-UHFFFAOYSA-N
Synonym 1-amino-4-hydroxyanthraquinone, disperse red 15, calcosyn pink b, celutate pink b, nacelan pink b, amacel pink b, celutate pink bn, celutate pink by, cibacete red 3b, disperse red 2s
PubChem CID 8323
IUPAC Name 1-amino-4-hydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)N

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 207.0°C to 209.0°C
Color Brown to Purple
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass Bottle
Quantity 5 g
Beilstein 14, 268
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 239.23
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-4-hydroxyanthraquinone, 96%
Show More Show Less
Hazard Category MUTAGEN
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Suspected of causing genetic defects.
May cause damage to organs.
May be fatal if swallowed and enters airways.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-163-0
RTECSNumber CB5600000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.