Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminoanthraquinone 98.0+%, TCI America™
SDP

Catalog No. A059025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A059025G 25 g
A0590500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A059025G Supplier TCI America Supplier No. A059025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82-45-1
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.231
MDL Number MFCD00001213
InChI Key KHUFHLFHOQVFGB-UHFFFAOYSA-N
Synonym 1-aminoanthraquinone, diazo fast red al, anthraquinone, 1-amino, 1-amino anthraquinone, 1-aminoanthrachinon, 1-amino-anthraquinone, 9,10-anthracenedione, 1-amino, alpha-aminoanthraquinone, 1-amino-9,10-anthraquinone, 1-amino-9,10-anthracenedione
PubChem CID 6710
IUPAC Name 1-aminoanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)N

Specifications

Melting Point 257°C
Quantity 25 g
UN Number 3077
Formula Weight 223.23
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminoanthraquinone
EINECSNumber (4)-0706
RTECSNumber CB5075000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.