Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminocyclohexanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. A1068100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1068100G 100 g
A106825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1068100G Supplier TCI America Supplier No. A1068100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2756-85-6
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00001487
InChI Key WOXWUZCRWJWTRT-UHFFFAOYSA-N
Synonym 1-aminocyclohexanecarboxylic acid, homocycloleucine, 1-amino-1-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-amino, alpha-aminocyclohexanecarboxylic acid, alpha-aminocyclohexanecarboxylic aicd, unii-1se3hd391h, 1-amino-1-cyclohexane carboxylic acid, .alpha.-aminocyclohexanecarboxylic acid, .alpha.-aminocyclohexanecarboxylic aicd
PubChem CID 1366
ChEBI CHEBI:86534
IUPAC Name 1-aminocyclohexane-1-carboxylic acid
SMILES NC1(CCCCC1)C(O)=O

Specifications

Color White
Quantity 100 g
Formula Weight 143.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminocyclohexanecarboxylic Acid
RTECSNumber GU8393000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.