Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminocyclopropane-1-Carboxylic Acid, MP Biomedicals™

Catalog No. icn15034880
Click to view available options
Quantity:
100 mg
250 mg
1 g

Chemical Identifiers

CAS 22059-21-8
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00009944
InChI Key PAJPWUMXBYXFCZ-UHFFFAOYSA-N
IUPAC Name 1-aminocyclopropane-1-carboxylic acid
SMILES NC1(CC1)C(O)=O

Specifications

Color White
Quantity 100 mg
Synonym piracetam, nootropil, 2-2-oxopyrrolidin-1-yl acetamide, 2-oxo-1-pyrrolidineacetamide, nootropyl, pyracetam, normabrain, gabacet, pyramem, 1-pyrrolidineacetamide, 2-oxo
PubChem CID 4843
Formula Weight 101.1
Physical Form Solid
Chemical Name or Material 1-Aminocyclopropane-1-Carboxylic Acid
Recommended Storage Store at 0°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.