Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminopyridinium Iodide 98.0+%, TCI America™
SDP

Catalog No. A062925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A062925G 25 g
A06295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A062925G Supplier TCI America Supplier No. A062925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6295-87-0
Molecular Formula C5H7IN2
Molecular Weight (g/mol) 222.03
MDL Number MFCD00051893
InChI Key NDRLPYIMWROJBG-UHFFFAOYSA-M
Synonym 1-aminopyridinium iodide, 1-aminopyridin-1-ium iodide, n-aminopyridinium iodide, pyridinium, 1-amino-, iodide, 1-aminopyridiniumiodide, pyridylamine, iodide, aminopyridinium iodide, amino-pyridinium iodide, l-aminopyridinium iodide, acmc-209nay
PubChem CID 2733704
IUPAC Name 1-aminopyridin-1-ium iodide
SMILES [I-].N[N+]1=CC=CC=C1

Specifications

Quantity 25 g
Formula Weight 222.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminopyridinium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.