Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aza-18-crown 6-Ether 98.0+%, TCI America™
SDP

Catalog No. p-7117372
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 33941-15-0
Molecular Formula C12H25NO5
Molecular Weight (g/mol) 263.334
MDL Number MFCD00075466
InChI Key NBXKUSNBCPPKRA-UHFFFAOYSA-N
PubChem CID 118578
IUPAC Name 1,4,7,10,13-pentaoxa-16-azacyclooctadecane
SMILES C1COCCOCCOCCOCCOCCN1

Specifications

Melting Point 51°C
Quantity 1 g
Synonym 1-aza-18-crown-6, 1-aza-18-crown6-ether, aza-18-crown-6, acmc-209tw0, 1-aza-18-crown 6-ether, 1-aza-18-crown nt, 1-aza-4,7,10,13,16-pentaoxacyclooctadecane
Formula Weight 263.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aza-18-crown 6-Ether
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.