Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzo[b]thiophen-3-yl-2-bromoethan-1-one, 97%, Thermo Scientific™

Catalog No. p-4510057
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 26167-45-3
Molecular Formula C10H7BrOS
Molecular Weight (g/mol) 255.129
InChI Key CKHWNGWAHFLCTJ-UHFFFAOYSA-N
Synonym 1-benzo b thiophen-3-yl-2-bromoethanone, 1-benzo b thiophen-3-yl-2-bromoethan-1-one, 3-bromoacetyl benzothiophene, 1-1-benzothiophen-3-yl-2-bromoethan-1-one, 3-bromoacetyl benzo b thiophene, 1-1-benzothiophen-3-yl-2-bromoethanone, 1-1-benzothiophen-3-yl-2-bromo-1-ethanone, 3-2-bromoacetyl benzo b thiophen, 1-benzothiophen-3-yl-2-bromo-ethanone, ethanone,1-benzo b thien-3-yl-2-bromo
PubChem CID 2776341
IUPAC Name 1-(1-benzothiophen-3-yl)-2-bromoethanone
SMILES C1=CC=C2C(=C1)C(=CS2)C(=O)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 255.13
Percent Purity 97%
Chemical Name or Material 1-Benzo[b]thiophen-3-yl-2-bromoethan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.