Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzothiophen-2-ylboronic acid, 97%, Thermo Scientific™

Catalog No. CC12212DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 98437-23-1
Molecular Formula C8H7BO2S
Molecular Weight (g/mol) 178.01
MDL Number MFCD01075674
InChI Key YNCYPMUJDDXIRH-UHFFFAOYSA-N
Synonym 2-benzothienylboronic acid, benzo b thiophen-2-ylboronic acid, benzo b thiophene-2-boronic acid, benzothiophene-2-boronic acid, 1-benzothien-2-ylboronic acid, thianaphthene-2-boronic acid, benzo b thiophene-2-ylboronic acid, 2-benzothiopheneboronic acid, benzothiophen-2-ylboronic acid
PubChem CID 2359
IUPAC Name (1-benzothiophen-2-yl)boronic acid
SMILES OB(O)C1=CC2=CC=CC=C2S1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.02
Percent Purity 97%
Chemical Name or Material 1-Benzothiophen-2-ylboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.