Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzothiophene-3-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC12304DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 5381-20-4
Molecular Formula C9H6OS
Molecular Weight (g/mol) 162.21
MDL Number MFCD00052376
InChI Key WDJLPQCBTBZTRH-UHFFFAOYSA-N
Synonym benzo b thiophene-3-carbaldehyde, benzo b thiophene-3-carboxaldehyde, thianaphthene-3-carboxaldehyde, 3-formylbenzo b thiophene, benzothiophene-3-carbaldehyde, 1-benzothiophen-3-carbaldehyd, pubchem13543, acmc-1apd4, thianapthene-3-carboxaldehyde, ksc274c6b
PubChem CID 227328
IUPAC Name 1-benzothiophene-3-carbaldehyde
SMILES O=CC1=CSC2=CC=CC=C12

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 162.21
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-3-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.