Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzoylacetone, 98%

Catalog No. AC105690010
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle

CAS: 93-91-4 | C10H10O2 | 162.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-91-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00008786
InChI Key CVBUKMMMRLOKQR-UHFFFAOYSA-N
Synonym benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton
PubChem CID 7166
IUPAC Name 1-phenylbutane-1,3-dione
SMILES CC(=O)CC(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 54°C to 58°C
Color Yellow
Density 1.0900g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5COCH2COCH3
Quantity 1 kg
Beilstein 07,68
Specific Gravity 1.09
Solubility Information Solubility in water: insoluble
Formula Weight 162.19
Percent Purity 98%
Physical Form Crystalline Flakes or Crystalline Powder
Chemical Name or Material 1-Benzoylacetone, 98%
Show More Show Less
EINECSNumber 202-286-4
RTECSNumber EK3540200
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.