Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-2-(methylsulfanyl)-1H-imidazole-5-carbaldehyde, 97%, Thermo Scientific™

Catalog No. cc28904da
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 479400-30-1
Molecular Formula C12H12N2OS
Molecular Weight (g/mol) 232.301
MDL Number MFCD04974046
InChI Key FELACOPTUQTZRP-UHFFFAOYSA-N
Synonym 1-benzyl-2-methylsulfanyl-1h-imidazole-5-carbaldehyde, 3-benzyl-2-methylsulfanyl imidazole-4-carbaldehyde, 2-methylthio-1-benzylimidazole-5-carbaldehyde, 1-benzyl-2-methylthio-1h-imidazole-5-carbaldehyde, 2-methylthio-3-benzyl-3h-imidazole-4-carbaldehyde
PubChem CID 343414
IUPAC Name 3-benzyl-2-methylsulfanylimidazole-4-carbaldehyde
SMILES CSC1=NC=C(N1CC2=CC=CC=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 232.31
Percent Purity 97%
Chemical Name or Material 1-Benzyl-2-(methylsulfanyl)-1H-imidazole-5-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.