Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-2-pyrrolidinone, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
5291-77-0
C11H13NO
175.23
MFCD00003195
LVUQCTGSDJLWCE-UHFFFAOYSA-N
1-benzyl-2-pyrrolidinone,n-benzyl-2-pyrrolidinone,n-benzyl-2-pyrrolidone,1-benzyl-2-pyrrolidone,n-benzylpyrrolidin-2-one,2-pyrrolidinone, 1-phenylmethyl,n-benzylpyrrolidone,1-benzylpyrrolidone,n-benzylpyrrolidinone,acmc-1aste
79176
1-benzylpyrrolidin-2-one
C1CC(=O)N(C1)CC2=CC=CC=C2
5291-77-0
C11H13NO
175.23
MFCD00003195
LVUQCTGSDJLWCE-UHFFFAOYSA-N
1-benzyl-2-pyrrolidinone,n-benzyl-2-pyrrolidinone,n-benzyl-2-pyrrolidone,1-benzyl-2-pyrrolidone,n-benzylpyrrolidin-2-one,2-pyrrolidinone, 1-phenylmethyl,n-benzylpyrrolidone,1-benzylpyrrolidone,n-benzylpyrrolidinone,acmc-1aste
79176
1-benzylpyrrolidin-2-one
C1CC(=O)N(C1)CC2=CC=CC=C2
Packaging Glass bottle
Flash Point >112°C
Density 1.0900g/mL
Specific Gravity 1.09
Refractive Index 1.5500 to 1.553
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 100g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 175.23
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-Benzyl-2-pyrrolidinone
Show More Show Less

EINECSNumber : 226-131-5