Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-2-pyrrolidinone, 97%

Catalog No. p-3777424
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 5291-77-0 | C11H13NO | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5291-77-0
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD00003195
InChI Key LVUQCTGSDJLWCE-UHFFFAOYSA-N
Synonym 1-benzyl-2-pyrrolidinone, n-benzyl-2-pyrrolidinone, n-benzyl-2-pyrrolidone, 1-benzyl-2-pyrrolidone, n-benzylpyrrolidin-2-one, 2-pyrrolidinone, 1-phenylmethyl, n-benzylpyrrolidone, 1-benzylpyrrolidone, n-benzylpyrrolidinone, acmc-1aste
PubChem CID 79176
IUPAC Name 1-benzylpyrrolidin-2-one
SMILES C1CC(=O)N(C1)CC2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0900g/mL
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5500 to 1.553
Quantity 25 g
Specific Gravity 1.09
Formula Weight 175.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Benzyl-2-pyrrolidinone
Show More Show Less
EINECSNumber 226-131-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.