Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-piperidone hydrate hydrochloride, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
50606-58-1
HCl·xH2O
225.72
MFCD00150096
OVWSFXNSJDMRPV-UHFFFAOYSA-N
1-benzyl-3-piperidone hydrochloride,1-benzylpiperidin-3-one hydrochloride,1-benzyl-3-piperidinone hydrochloride,1-benzyl-3-piperidone hydrate hydrochloride,1-benzylpiperidin-3-one hcl,1-benzyl-3-piperidone hcl hydrate,3-piperidinone, 1-phenylmethyl-, hydrochloride,1-benzyl-3-piperidinone hydrate hydrochloride,n-benzyl-3-piperidone hydrochloride,pubchem7726
3084924
1-benzylpiperidin-3-one;hydrochloride
C1CC(=O)CN(C1)CC2=CC=CC=C2.Cl
50606-58-1
HCl·xH2O
225.72
MFCD00150096
OVWSFXNSJDMRPV-UHFFFAOYSA-N
1-benzyl-3-piperidone hydrochloride,1-benzylpiperidin-3-one hydrochloride,1-benzyl-3-piperidinone hydrochloride,1-benzyl-3-piperidone hydrate hydrochloride,1-benzylpiperidin-3-one hcl,1-benzyl-3-piperidone hcl hydrate,3-piperidinone, 1-phenylmethyl-, hydrochloride,1-benzyl-3-piperidinone hydrate hydrochloride,n-benzyl-3-piperidone hydrochloride,pubchem7726
3084924
1-benzylpiperidin-3-one;hydrochloride
C1CC(=O)CN(C1)CC2=CC=CC=C2.Cl
Melting Point 170°C to 176°C
Water 8% max. (K.F.)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Color Brown to Cream
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (on free base) (GC)
Solubility Information Solubility in water: soluble
Formula Weight 225.72
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 1-Benzyl-3-piperidone hydrate hydrochloride, 95%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 256-646-0

TSCA : TSCA