Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-piperidone Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. B34195G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 50606-58-1
Molecular Formula C12H16ClNO
Molecular Weight (g/mol) 225.72
MDL Number MFCD00150096
InChI Key OVWSFXNSJDMRPV-UHFFFAOYSA-N
Synonym 1-benzyl-3-piperidone hydrochloride, 1-benzylpiperidin-3-one hydrochloride, 1-benzyl-3-piperidinone hydrochloride, 1-benzyl-3-piperidone hydrate hydrochloride, 1-benzylpiperidin-3-one hcl, 1-benzyl-3-piperidone hcl hydrate, 3-piperidinone, 1-phenylmethyl-, hydrochloride, 1-benzyl-3-piperidinone hydrate hydrochloride, n-benzyl-3-piperidone hydrochloride, pubchem7726
PubChem CID 3084924
IUPAC Name 1-benzylpiperidin-3-one hydrochloride
SMILES Cl.O=C1CCCN(CC2=CC=CC=C2)C1

Specifications

Melting Point 171°C
Boiling Point 91°C
Quantity 5 g
Formula Weight 225.72
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-3-piperidone Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.