Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-pyrrolidone 98.0+%, TCI America™
SDP

Catalog No. B36065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36065G 5 g
B360625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36065G Supplier TCI America Supplier No. B36065G

Chemical Identifiers

CAS 775-16-6
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00005342
InChI Key DHGMDHQNUNRMIN-UHFFFAOYSA-N
Synonym 1-benzyl-3-pyrrolidinone, n-benzyl-3-pyrrolidinone, 1-phenylmethyl-3-pyrrolidinone, 1-benzyl-3-pyrrolidone, 1-benzyl-pyrrolidin-3-one, 1-benzyl pyrrolidin-3-one, 3-pyrrolidinone, 1-phenylmethyl, benzyl-3-pyrrolidinone, pubchem10007, n-benzylpyrrolid-3-one
PubChem CID 69890
IUPAC Name 1-benzylpyrrolidin-3-one
SMILES C1CN(CC1=O)CC2=CC=CC=C2

Specifications

Boiling Point 109°C
Quantity 5 g
Formula Weight 175.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-pyrrolidone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.