Learn More
1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride, 99%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS | 71258-18-9 |
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Molecular Formula | C19H21ClN2 |
| Quantity | 100g |
| MDL Number | MFCD00035361 |
| Synonym | 1-benzyl-4-cyano-4-phenylpiperidine hydrochloride, 1-benzyl-4-phenylpiperidine-4-carbonitrile hydrochloride, 1-benzyl-4-cyano-4-phenylpiperidine hcl, 1-n-benzyl-4-cyano-4-phenylpiperidine hydrochloride, n-benzyl-4-cyano-4-phenylpiperidine hydrochloride, 4-phenyl-1-benzylpiperidine-4-carbonitrile, chloride, 4-phenyl-1-phenylmethyl-4-piperidinecarbonitrile hydrochloride, pubchem13098, acmc-1bfv8, c19h20n2.clh |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: Harmful in contact with skin. Harmful if inhaled. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.