Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-4-cyano-4-phenylpiperidine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B439325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B439325G 25 g
B43935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B439325G Supplier TCI America Supplier No. B439325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71258-18-9
Molecular Formula C19H21ClN2
Molecular Weight (g/mol) 312.841
MDL Number MFCD00035361
InChI Key SPXFNRMZPRHYRH-UHFFFAOYSA-N
Synonym 1-benzyl-4-cyano-4-phenylpiperidine hydrochloride, 1-benzyl-4-phenylpiperidine-4-carbonitrile hydrochloride, 1-benzyl-4-cyano-4-phenylpiperidine hcl, 1-n-benzyl-4-cyano-4-phenylpiperidine hydrochloride, n-benzyl-4-cyano-4-phenylpiperidine hydrochloride, 4-phenyl-1-benzylpiperidine-4-carbonitrile, chloride, 4-phenyl-1-phenylmethyl-4-piperidinecarbonitrile hydrochloride, pubchem13098, acmc-1bfv8, c19h20n2.clh
PubChem CID 12652062
IUPAC Name 1-benzyl-4-phenylpiperidine-4-carbonitrile;hydrochloride
SMILES C1CN(CCC1(C#N)C2=CC=CC=C2)CC3=CC=CC=C3.Cl

Specifications

Melting Point 263°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 312.84
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-4-cyano-4-phenylpiperidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.