Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-4-piperidinecarboxaldehyde, 97%

Catalog No. AC436590010
Click to view available options
:
1g

CAS: 22065-85-6 | C13H17NO | 203.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 22065-85-6
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.026g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C13H17NO
Quantity 1 g
Synonym 1-benzyl-4-piperidinecarboxaldehyde, n-benzylpiperidine-4-carboxaldehyde, 1-benzyl-4-formylpiperidine, 1-benzylpiperidine-4-carboxaldehyde, 1-benzy-4-piperidine carboxyaldehyde, 1-benzyl-4-piperidinecarbaldehyde, 1-benzyl-piperidine-4-carbaldehyde, n-benzyl piperidine-4-carboxaldehyde, 4-piperidinecarboxaldehyde, 1-phenylmethyl, n-benzyl-4-formylpiperidine
InChI Key SGIBOXBBPQRZDM-UHFFFAOYSA-N
SMILES C1CN(CCC1C=O)CC2=CC=CC=C2
IUPAC Name 1-benzylpiperidine-4-carbaldehyde
Molecular Weight (g/mol) 203.285
PubChem CID 89584
Formula Weight 203.28
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 1-Benzyl-4-piperidinecarboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.