Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Benzyl-4-piperidone, 98+%
Click to view available options
Quantity:
25 g
100 g
500 g
Description
1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.
Solubility
Soluble in water (12 g/100ml at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.
Solubility
Soluble in water (12 g/100ml at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
Density | 1.059 |
Boiling Point | 248°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.5399 |
Quantity | 500 g |
Beilstein | 128556 |
Solubility Information | Soluble in water (12g/100ml at 20°C). |
Formula Weight | 189.26 |
Percent Purity | ≥98% |
Chemical Name or Material | 1-Benzyl-4-piperidone |
Safety and Handling
Hazard Category | H227-H302-H315-H317-H319 |
Hazard Statement | GHS H Statement H301-H317 Toxic if swallowed. May cause an allergic skin reaction. |
Precautionary Statement | P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P370+P378q-P501c |
EINECSNumber | 222-782-4 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction