Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-4-piperidone 98.0+%, TCI America™
SDP

Catalog No. b102725ml
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 3612-20-2
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
MDL Number MFCD00006192
InChI Key SJZKULRDWHPHGG-UHFFFAOYSA-N
Synonym 1-benzyl-4-piperidone, n-benzyl-4-piperidone, 4-piperidinone, 1-phenylmethyl, 1-benzyl-4-piperidinone, 1-benzylpiperidone, 1-benzyl-4-oxopiperidine, 1-benzyl-piperidin-4-one, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzylpiperidin-4-one
PubChem CID 19220
IUPAC Name 1-benzylpiperidin-4-one
SMILES C1CN(CCC1=O)CC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 158°C
Quantity 25 mL
Formula Weight 189.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-4-piperidone
EINECSNumber (5)-3734
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.