Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzylimidazole, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
4238-71-5
C10H10N2
158.2
MFCD00005296
KKKDZZRICRFGSD-UHFFFAOYSA-N
1-benzyl-1h-imidazole,n-benzylimidazole,imidazole, 1-benzyl,benzylimidazole,1h-imidazole, 1-phenylmethyl,1-benzyl imidazole,ccris 5821,chembl14192,1bn,1-benzylimdazole
77918
1-benzylimidazole
C1=CC=C(C=C1)CN2C=CN=C2
4238-71-5
C10H10N2
158.2
MFCD00005296
KKKDZZRICRFGSD-UHFFFAOYSA-N
1-benzyl-1h-imidazole,n-benzylimidazole,imidazole, 1-benzyl,benzylimidazole,1h-imidazole, 1-phenylmethyl,1-benzyl imidazole,ccris 5821,chembl14192,1bn,1-benzylimdazole
77918
1-benzylimidazole
C1=CC=C(C=C1)CN2C=CN=C2
Packaging Glass bottle
Boiling Point 310.0°C
Quantity 100g
Infrared Spectrum Authentic
Color Brown
Melting Point 68.0°C to 71.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 23, 48
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol and acetone
Formula Weight 158.2
Physical Form Crystals
Percent Purity 99%
Chemical Name or Material 1-Benzylimidazole
Show More Show Less