Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzylimidazole 98.0+%, TCI America™
SDP

Catalog No. B338725G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 4238-71-5
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00005296
InChI Key KKKDZZRICRFGSD-UHFFFAOYSA-N
Synonym 1-benzyl-1h-imidazole, n-benzylimidazole, imidazole, 1-benzyl, benzylimidazole, 1h-imidazole, 1-phenylmethyl, 1-benzyl imidazole, ccris 5821, chembl14192, 1bn, 1-benzylimdazole
PubChem CID 77918
IUPAC Name 1-benzyl-1H-imidazole
SMILES C(N1C=CN=C1)C1=CC=CC=C1

Specifications

Melting Point 73°C
Boiling Point 122°C
Quantity 25 g
Formula Weight 158.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzylimidazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.