Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyloxy-2-methyl-3-nitrobenzene, 98%, Thermo Scientific™

Catalog No. AAH5589906
Click to view available options
Quantity:
1 g
5 g
25 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20876-37-3
Molecular Formula C14H13NO3
Molecular Weight (g/mol) 243.262
MDL Number MFCD00100652
InChI Key PBSZHNXXFIYDBU-UHFFFAOYSA-N
Synonym 1-benzyloxy-2-methyl-3-nitrobenzene, 2-benzyloxy-6-nitrotoluene, 6-benzyloxy-2-nitrotoluene, benzene, 2-methyl-1-nitro-3-phenylmethoxy, acmc-1cjof, maybridge1_002032, ksc496g7p, benzyl 3-nitro-o-tolyl ether, 1-benzyloxy-2-methyl-3-nitrobenzene #
PubChem CID 292648
IUPAC Name 2-methyl-1-nitro-3-phenylmethoxybenzene
SMILES CC1=C(C=CC=C1OCC2=CC=CC=C2)[N+](=O)[O-]

Specifications

Melting Point 60°C to 63°C
Color Brown to Yellow
Quantity 5 g
Formula Weight 243.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Benzyloxy-2-methyl-3-nitrobenzene
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.