Learn More
1-Benzyloxycarbonyl-3-piperidone, 97+%
Description
1-Benzyloxycarbonyl-3-piperidone finds its application in the synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Benzyloxycarbonyl-3-piperidone finds its application in the synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.
Solubility
Slightly soluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications
Specifications
| Quantity | 5 g |
| Beilstein | 1464624 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 233.27 |
| Percent Purity | ≥97% |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Benzyloxycarbonyl-3-piperidone |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.