Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Biotinyl-3 6-Dioxa-8-Octaneamine Hydrochloride

Catalog No. 91446050MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-446-050MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-446-050MG Supplier Sigma Aldrich Supplier No. 91446050MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Biotinyl-3,6-dioxa-8-octaneamine hydrochloride is a versatile biotinylated linker that can be incorporated into chemical tools via its terminal amino group. Labeling materials or proteins with biotin provides a means to enrich and capture targets from biological systems.Efficient Innate Immune Killing of Cancer Cells Triggered by Cell Surface Anchoring of Multivalent Antibody-Recruiting PolymersCaged Cyclopropenes with Improved Tetrazine Ligation KineticsA Unified Framework for the Incorporation of Bioorthogonal Compound Exposure Probes within Biological CompartmentsA Chemical Probe for Protein CrotonylationOne-Step Selective Exoenzymatic Labeling (SEEL) Strategy for the Biotinylation and Identification of Glycoproteins of Living Cells

Specifications

Molecular Formula C53H45NO5
Linear Formula C25H37FN4O5
Quantity 100 mg
Synonym 2-(2-Ethylhexyl)-6,8,11-tris(4-methoxyphenyl)-1H-peryleno[1,12-efg]isoindole-1,3(2H)-dione; BPI-p-OMePh
Molecular Weight (g/mol) 775.93 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.