Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[Bis(4-fluorophenyl)methyl]piperazine 97.0+%, TCI America™
SDP

Catalog No. B276425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B276425G 25 g
B27645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B276425G Supplier TCI America Supplier No. B276425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27469-60-9
Molecular Formula C17H18F2N2
Molecular Weight (g/mol) 288.342
InChI Key TTXIFFYPVGWLSE-UHFFFAOYSA-N
Synonym 1-bis 4-fluorophenyl methyl piperazine, 1-bis-4-fluorophenyl methyl piperazine, 1-4,4'-difluorobenzhydryl piperazine, 4,4'-difluorobenzhydrylpiperazine, 1-bis 4-fluorophenyl methylpiperazine, 1 bis 4-fluorophenyl methyl-piperazine, piperazine, 1-bis 4-fluorophenyl methyl, 4,4'-difluorobenzhydryl piperazine
PubChem CID 152932
IUPAC Name 1-[bis(4-fluorophenyl)methyl]piperazine
SMILES C1CN(CCN1)C(C2=CC=C(C=C2)F)C3=CC=C(C=C3)F

Specifications

Melting Point 93°C
Color White-Yellow
Boiling Point 130°C
Quantity 25 g
UN Number 2811
Formula Weight 288.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-[Bis(4-fluorophenyl)methyl]piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.