Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Boc-2-trifluoromethyl-4-piperidone, 95%

Click to view available options
Quantity:
250mg
1g
1245648-32-1
C11H16F3NO3
267.248
MFCD07367733
RSFRBHIRISNXIK-UHFFFAOYSA-N
tert-butyl 4-oxo-2-trifluoromethyl piperidine-1-carboxylate,1-boc-2-trifluoromethyl-piperidin-4-one,1-boc-2-trifluromethylpiperidin-4-one,1-boc-2-trifluoromethylpiper idin-4-one,t-butyl 4-oxo-2-trifluoromethyl piperidine-1-carboxylate,tert-butyl4-oxo-2-trifluoromethyl piperidine-1-carboxylate,tert-butyl 4-oxo-2-trifluoromethyl ;piperidine-1-carboxylate,1-boc-2-trifluromethyl-piperidin-4-one;4-oxo-2-trifluoromethyl-piperidine-1-carboxylic acid tert-butyl ester
71607278
tert-butyl 4-oxo-2-(trifluoromethyl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(=O)CC1C(F)(F)F
1245648-32-1
C11H16F3NO3
267.248
MFCD07367733
RSFRBHIRISNXIK-UHFFFAOYSA-N
tert-butyl 4-oxo-2-trifluoromethyl piperidine-1-carboxylate,1-boc-2-trifluoromethyl-piperidin-4-one,1-boc-2-trifluromethylpiperidin-4-one,1-boc-2-trifluoromethylpiper idin-4-one,t-butyl 4-oxo-2-trifluoromethyl piperidine-1-carboxylate,tert-butyl4-oxo-2-trifluoromethyl piperidine-1-carboxylate,tert-butyl 4-oxo-2-trifluoromethyl ;piperidine-1-carboxylate,1-boc-2-trifluromethyl-piperidin-4-one;4-oxo-2-trifluoromethyl-piperidine-1-carboxylic acid tert-butyl ester
71607278
tert-butyl 4-oxo-2-(trifluoromethyl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(=O)CC1C(F)(F)F
Quantity 250mg
Formula Weight 267.25
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 1-Boc-2-trifluoromethyl-4-piperidone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning