Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-BOC-3-aminoazetidine, 95%

Catalog No. AC451180010
Click to view available options
Quantity:
1 g
5 g

CAS: 193269-78-2 | C8H16N2O2 | 172.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193269-78-2
Molecular Formula C8H16N2O2
Molecular Weight (g/mol) 172.23
InChI Key WPGLRFGDZJSQGI-UHFFFAOYSA-N
Synonym 1-boc-3-amino azetidine, 1-boc-3-aminoazetidine, 3-amino-1-n-boc-azetidine, 1-n-boc-3-aminoazetidine, 3-amino-1-boc-azetidine, 3-amino-1-n-boc azetidine, tert-butyl 3-aminoazetidinecarboxylate, 1-azetidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-aminoazetidine, 3-amino-1-tert-butoxycarbonyl azetidine
PubChem CID 1516507
IUPAC Name tert-butyl 3-aminoazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)N

Specifications

Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 172.23
Percent Purity 95%
Chemical Name or Material 1-BOC-3-aminoazetidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.