Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-BOC-4-Piperidinecarboxaldehyde, 95%, Thermo Scientific™

Catalog No. AC441755000
Click to view available options
Quantity:
500 mg
Packaging:
Glass Bottle

Chemical Identifiers

CAS 137076-22-3
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.28
MDL Number MFCD02179019
InChI Key JYUQEWCJWDGCRX-UHFFFAOYSA-N
Synonym 1-boc-4-piperidinecarboxaldehyde, 1-boc-4-formylpiperidine, 1-tert-butoxycarbonyl-4-piperidinecarboxaldehyde, 1-boc-piperidine-4-carboxaldehyde, n-boc-4-formylpiperidine, n-boc-4-piperidinecarboxaldehyde, 1-tert-butoxycarbonyl-4-formylpiperidine, 4-formyl-piperidine-1-carboxylic acid tert-butyl ester, n-boc-piperidine-4-carbaldehyde, 4-formylpiperidine-1-carboxylic acid tert-butyl ester
PubChem CID 1514430
IUPAC Name tert-butyl 4-formylpiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C=O

Specifications

Melting Point 31.0°C to 34.0°C
Boiling Point 120.0°C (2.0 mmHg)
Flash Point 110°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 500 mg
Formula Weight 213.28
Percent Purity 95%
Chemical Name or Material 1-BOC-4-Piperidinecarboxaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minu

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.