Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-azetidine-3-carboxaldehyde, 97%

Catalog No. AAH5279406
Click to view available options
Quantity:
250 mg
1 g
5 g

Catalog No. AAH5279406

Supplier: Thermo Scientific Chemicals H5279406

Only null left
Add to Cart

CAS: 177947-96-5 | C9H15NO3 | 185.223 g/mol

1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.

Notes
Air sensitive. Store in a cool place. Incompatible with water and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177947-96-5
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD06656142
InChI Key JVQOZRRUGOADSU-UHFFFAOYSA-N
Synonym 1-boc-3-azetidinecarboxaldehyde, 1-boc-3-formylazetidine, 1-boc-azetidine-3-carboxaldehyde, azetidine-3-carboxaldehyde, 1-azetidinecarboxylic acid, 3-formyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl azetidine-3-carboxaldehyde, 3-formylazetidine-1-carboxylic acid tert-butylester, pubchem10136, 1-boc-3-azetidinecarbaldehyde, 1-boc-3-azetidine carboxaldehyde
PubChem CID 10726182
IUPAC Name tert-butyl 3-formylazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)C=O

Specifications

Boiling Point 77°C to 82°C (2mmHg)
Quantity 5 g
Formula Weight 185.22
Sensitivity Air sensitive
Percent Purity 97%
Chemical Name or Material 1-Boc-azetidine-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.