Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-N-Fmoc-D-tryptophan, 98%

Catalog No. p-7041461
Click to view available options
Quantity:
1 g
5 g

CAS: 163619-04-3 | C31H30N2O6 | 526.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163619-04-3
Molecular Formula C31H30N2O6
Molecular Weight (g/mol) 526.59
MDL Number MFCD00153367
InChI Key ADOHASQZJSJZBT-AREMUKBSSA-N
Synonym fmoc-d-trp boc-oh, n, a-fmoc-n in-boc-d-tryptophan, fmoc-d-trp boc, ambotzfaa1339, pubchem10051, 1-boc-fmoc-d-tryptophan, 1-boc-n-fmoc-d-tryptophan, 2r-3-1-tert-butoxycarbonylindol-3-yl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, nalpha-fmoc-n in-boc-d-tryptophan, fmoc-d-trp boc-oh hplc
PubChem CID 16213150
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]propanoic acid
SMILES CC(C)(C)OC(=O)N1C=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 7062970
Formula Weight 526.59
Percent Purity 98%
Chemical Name or Material 1-Boc-N-Fmoc-D-tryptophan
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.