Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-pyrrole-2-carboxaldehyde, 98%, Thermo Scientific™

Catalog No. AAH3255306
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAH3255306

Supplier: Thermo Scientific Chemicals H3255306

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161282-57-1
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD03086220
InChI Key NSHLHMBABZKJHP-UHFFFAOYSA-N
Synonym tert-butyl 2-formyl-1h-pyrrole-1-carboxylate, n-boc-pyrrole-2-carboxaldehyde, 2-formyl-pyrrole-1-carboxylic acid tert-butyl ester, acmc-20ak5v, n-boc-pyrrole-2-carbaldehyde, 1-boc-pyrrole-2-carboxaldehyde, 1-t-butoxycarbonyl pyrrole-2-carbaldehyde, tert-butyl 2-methanoylpyrrole-1-carboxylate
PubChem CID 5057005
IUPAC Name tert-butyl 2-formylpyrrole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C=CC=C1C=O

Specifications

Melting Point 49°C to 52°C
Flash Point 107.0°C (225°F)
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 195.22
Percent Purity 98%
Chemical Name or Material 1-Boc-pyrrole-2-carboxaldehyde
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.