Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,2-dimethoxypropane, 98%

Catalog No. AAA1113918
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 126-38-5 | C5H11BrO2 | 183.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-38-5
Molecular Formula C5H11BrO2
Molecular Weight (g/mol) 183.045
MDL Number MFCD00000208
InChI Key SGTITUFGCGGICE-UHFFFAOYSA-N
Synonym propane,1-bromo-2,2-dimethoxy, propane, 1-bromo-2,2-dimethoxy, bromoacetone dimethyl acetal, bromo-2,2dimethoxypropane, bromo-2,2-dimethoxypropane, 1-bromo-2,2-dimethoxypropan, 1-bromo-2,2-dimethoxyprpopane, 1-bromo-2,2-dimethoxy propane, 1-bromo-2,2-dimethoxy-propane, bromoacetone dimethyl ketal
PubChem CID 67175
IUPAC Name 1-bromo-2,2-dimethoxypropane
SMILES CC(CBr)(OC)OC

Specifications

Density 1.354
Boiling Point 83°C to 87°C (80 mmHg)
Flash Point 40°C (104°F)
Refractive Index 1.447
Quantity 50 g
UN Number UN3271
Beilstein 2036095
Sensitivity Moisture Sensitive
Formula Weight 183.05
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,2-dimethoxypropane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 204-784-7
TSCA No

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.