Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,3-dichlorobenzene 99.0+%, TCI America™
SDP

Catalog No. B098025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B098025G 25 g
B09805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B098025G Supplier TCI America Supplier No. B098025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56961-77-4
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.89
MDL Number MFCD00000536
InChI Key HVKCZUVMQPUWSX-UHFFFAOYSA-N
Synonym 2,3-dichlorobromobenzene, benzene, 1-bromo-2,3-dichloro, benzene, bromodichloro, 2,3-dichloro-1-bromobenzene, 1-bromo-2,3-dichloro-benzene, attercop-chm at113305, bromodichlorobenzene, pubchem3607, monobromodichlorobenzene, acmc-209lv2
PubChem CID 42066
IUPAC Name 1-bromo-2,3-dichlorobenzene
SMILES ClC1=CC=CC(Br)=C1Cl

Specifications

Melting Point 60°C
Boiling Point 243°C
Quantity 25 g
Formula Weight 225.89
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2,3-dichlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.