Learn More
1-Bromo-2,4-dinitrobenzene, 98%
Description
1-Bromo-2,4-dinitrobenzene has been used in the preparation of 2,4-dinitrophenol via treatment with KO2-crown ether complex in benzene. It is used as substrate in protein determination and glutathione S-transferase (GST) assay of chicken and rat prostaglandin D2 synthase (PGDS).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Bromo-2,4-dinitrobenzene has been used in the preparation of 2,4-dinitrophenol via treatment with KO2-crown ether complex in benzene. It is used as substrate in protein determination and glutathione S-transferase (GST) assay of chicken and rat prostaglandin D2 synthase (PGDS).
Solubility
Soluble in water (partly miscible).
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 70°C to 73°C |
Quantity | 100 g |
UN Number | UN3459 |
Synonym | 2,4-dinitrobromobenzene, 4-bromo-1,3-dinitrobenzene, o,p-dinitrophenyl bromide, 2,4-dinitrophenyl bromide, dnbb, benzene, 1-bromo-2,4-dinitro, 2, 4-dinitrobromobenzene, 1-bromo-2,4-dinitrobenze, unii-4ms3ftw380, ccris 1801 |
Solubility Information | Soluble in water (partly miscible). |
Formula Weight | 247.01 |
Percent Purity | 98% |
Chemical Name or Material | 1-Bromo-2,4-dinitrobenzene |
Safety and Handling
Hazard Category | H301+H311+H331-H314 |
Hazard Statement | GHS H Statement H302-H312-H315-H319-H317-H335 Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: II |
EINECSNumber | 209-539-8 |
RTECSNumber | CY9021500 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.