Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-butyne, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3355-28-0
C4H5Br
132.99
MFCD00190233
LNNXOEHOXSYWLD-UHFFFAOYSA-N
1-bromo-2-butyne,2-butyne, 1-bromo,2-butynyl bromide,1-bromo-but-2-yne,bromo-2-butyne,1-bromo2-butyne,but-2-ynylbromide,l-bromo-2-butine,1-bromo-2-butine,4-bromo-2-butyne
2756862
1-bromobut-2-yne
CC#CCBr
3355-28-0
C4H5Br
132.99
MFCD00190233
LNNXOEHOXSYWLD-UHFFFAOYSA-N
1-bromo-2-butyne,2-butyne, 1-bromo,2-butynyl bromide,1-bromo-but-2-yne,bromo-2-butyne,1-bromo2-butyne,but-2-ynylbromide,l-bromo-2-butine,1-bromo-2-butine,4-bromo-2-butyne
2756862
1-bromobut-2-yne
CC#CCBr
Specific Gravity 1.473
Boiling Point 40.0°C to 41.0°C
Refractive Index 1.5070 to 1.5090 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Density 1.4730g/mL
Quantity 1g
Color Green-Yellow
Flash Point 36°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CCCH2Br
Formula Weight 132.99
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-butyne
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning