Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-ethylbenzene, 98%

Catalog No. AAB2344922
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1973-22-4 | C8H9Br | 185.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1973-22-4
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000077
InChI Key HVRUGFJYCAFAAN-UHFFFAOYSA-N
PubChem CID 16089
IUPAC Name 1-bromo-2-ethylbenzene
SMILES CCC1=CC=CC=C1Br

Specifications

Melting Point -68°C
Density 1.338
Boiling Point 199°C to 200°C
Flash Point 71°C (159°F)
Refractive Index 1.5485
Quantity 100 g
Beilstein 1929735
Synonym o-bromoethylbenzene, benzene, 1-bromo-2-ethyl, 2-ethylbromobenzene, 1-ethyl-2-bromobenzene, 2-bromo-1-ethylbenzene, 1-bromo-2-ethyl-benzene, benzene,1-bromo-2-ethyl, o-ethylbromobenzene, 2-bromophenyl ethane, bromo-2-ethylbenzene
Formula Weight 185.06
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-ethylbenzene
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 217-823-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.