Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-ethylbutane, 97%, Thermo Scientific Chemicals

Catalog No. AC305180250
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3814-34-4
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.08
MDL Number MFCD00000219
InChI Key KKGUMGWNFARLSL-UHFFFAOYSA-N
Synonym 1-bromo-2-ethylbutane, 3-bromomethyl pentane, 2-ethylbutyl bromide, pentane, 3-bromomethyl, 3-bromomethyl-pentane, butane, 1-bromo-2-ethyl, 2-ethylbutylbromide, bromo-2-ethylbutane, 2-ethyl butyl bromide, 2-ethyl-1-bromobutane
PubChem CID 77432
IUPAC Name 3-(bromomethyl)pentane
SMILES CCC(CC)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 143°C to 144°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4510 to 1.453
Linear Formula (C2H5)2CHCH2Br
Quantity 25 mL
Specific Gravity 1.17
Formula Weight 165.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-ethylbutane, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-302-6
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.