Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-methoxymethylnaphthalene 98.0+%, TCI America™
SDP

Catalog No. B117025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B117025G 25 g
B11705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B117025G Supplier TCI America Supplier No. B117025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64689-70-9
Molecular Formula C12H11BrO
Molecular Weight (g/mol) 251.123
MDL Number MFCD00059518
InChI Key ORYVAZPUJFACFR-UHFFFAOYSA-N
PubChem CID 12385765
IUPAC Name 1-bromo-2-(methoxymethyl)naphthalene
SMILES COCC1=C(C2=CC=CC=C2C=C1)Br

Specifications

Boiling Point 151°C
Quantity 25 g
Formula Weight 251.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-methoxymethylnaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.