Learn More
1-Bromo-2-methylpropane, 98+%
Description
1-Bromo-2-methylpropane is used as a synthetic reagent for the preparation of 1-ethyl-1,3-isobutylimidazolium bromide by reaction with 1-ethylimidazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Bromo-2-methylpropane is used as a synthetic reagent for the preparation of 1-ethyl-1,3-isobutylimidazolium bromide by reaction with 1-ethylimidazole.
Solubility
Slightly soluble in water. Miscible with alcohol, ether.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | -118°C |
| Density | 1.255 |
| Boiling Point | 90°C to 92°C |
| Flash Point | 18°C (64°F) |
| Refractive Index | 1.435 |
| Quantity | 1000 g |
| UN Number | UN2342 |
| Beilstein | 1730915 |
| Merck Index | 14,5135 |
| Solubility Information | Slightly soluble in water; Miscible with alcohol,ether. |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H319-H335 |
| Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BROMOMETHYLPROPANES |
| EINECSNumber | 201-141-2 |
| RTECSNumber | TX4140000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.